Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS19
Entrez Gene ID
102175432
Gene Name
ribosomal protein S19
Gene Aliases
-
Chromosome Location
Chr.18: 52255059 - 52263068 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692574.3 XP_005692631.1 5 - 6 919 99
XM_018062504.1 XP_017917993.1 5 - 6 589 99
GenBank mRNA JO452910.2 - 1 - 2 75 99
KA335100.1 - 1 - 2 129 99

View Full Product Details