Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A13
Entrez Gene ID
102175444
Gene Name
solute carrier family 22 member 13
Gene Aliases
-
Chromosome Location
Chr.22: 11601370 - 11614813 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066630.1 XP_017922119.1 2 - 3 489 108

View Full Product Details