Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUC20
Entrez Gene ID
102174698
Gene Name
mucin 20, cell surface associated
Gene Aliases
-
Chromosome Location
Chr.1: 70225886 - 70241267 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675240.3 XP_005675297.1 3 - 4 1760 64
GenBank mRNA JO428694.2 - 1 - 2 599 64
JO430144.2 - 1 - 2 599 64
KA348692.1 - 2 - 3 359 64

View Full Product Details