Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL9
Entrez Gene ID
102178087
Gene Name
myosin light chain 9
Gene Aliases
-
Chromosome Location
Chr.13: 65213310 - 65221684 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688518.3 XP_005688575.1 1 - 2 364 78

View Full Product Details