Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL9
Entrez Gene ID
102178087
Gene Name
myosin light chain 9
Gene Aliases
-
Chromosome Location
Chr.13: 65213310 - 65221684 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688518.3 XP_005688575.1 2 - 3 574 96
XM_018057813.1 XP_017913302.1 2 - 3 319 96

View Full Product Details