Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC38
Entrez Gene ID
102172178
Gene Name
leucine rich repeat containing 38
Gene Aliases
-
Chromosome Location
Chr.16: 52575587 - 52611539 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690878.3 XP_005690935.2 1 - 2 1287 58

View Full Product Details