Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLD4
Entrez Gene ID
102182284
Gene Name
DNA polymerase delta 4, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.29: 45638008 - 45639778 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699945.3 XP_005700002.1 1 - 2 303 115

View Full Product Details