Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP3
Entrez Gene ID
102179220
Gene Name
THAP domain containing 3
Gene Aliases
-
Chromosome Location
Chr.16: 45315306 - 45321301 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013970245.2 XP_013825699.2 2 - 3 2645 107
XM_018060414.1 XP_017915903.1 3 - 4 397 107
GenBank mRNA KA343453.1 - 2 - 3 883 107

View Full Product Details