Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHD2
Entrez Gene ID
102171359
Gene Name
EF-hand domain family member D2
Gene Aliases
-
Chromosome Location
Chr.16: 50547277 - 50565264 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060594.1 XP_017916083.1 2 - 3 535 61

View Full Product Details