Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAC3
Entrez Gene ID
102178569
Gene Name
Rac family small GTPase 3
Gene Aliases
-
Chromosome Location
Chr.19: 50249519 - 50254145 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065431.1 XP_017920920.1 5 - 6 719 79
XM_018065432.1 XP_017920921.1 5 - 6 1434 79
GenBank mRNA JO604644.2 - 1 - 2 128 79

View Full Product Details