Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAA80
Entrez Gene ID
102172493
Gene Name
N(alpha)-acetyltransferase 80, NatH catalytic subunit
Gene Aliases
NAT6
Chromosome Location
Chr.22: 50114479 - 50117169 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066625.1 XP_017922114.1 4 - 4 1069 121

View Full Product Details