Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTX4
Entrez Gene ID
102180364
Gene Name
pentraxin 4
Gene Aliases
-
Chromosome Location
Chr.25: 1077210 - 1080998 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039847.1 XP_017895336.1 2 - 3 773 70

View Full Product Details