Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TSC22D4
Entrez Gene ID
102190374
Gene Name
TSC22 domain family member 4
Gene Aliases
-
Chromosome Location
Chr.25: 36949261 - 36960220 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040776.1 XP_017896265.1 3 - 4 1960 71
GenBank mRNA JO392910.2 - 2 - 3 631 71
JO427808.1 - 2 - 3 703 71

View Full Product Details