Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARD6G
Entrez Gene ID
102187092
Gene Name
par-6 family cell polarity regulator gamma
Gene Aliases
-
Chromosome Location
Chr.24: 222160 - 298692 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039768.1 XP_017895257.1 2 - 3 543 78
GenBank mRNA KA353317.1 - 2 - 3 406 78

View Full Product Details