Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGAS
Entrez Gene ID
102187049
Gene Name
cyclic GMP-AMP synthase
Gene Aliases
MB21D1
Chromosome Location
Chr.9: 104206 - 129766 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013966327.2 XP_013821781.2 3 - 4 1157 106
GenBank mRNA JT716109.1 - 6 - 7 1037 106

View Full Product Details