Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGO2
Entrez Gene ID
102187354
Gene Name
shugoshin 2
Gene Aliases
-
Chromosome Location
Chr.2: 46839699 - 46883229 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061146.1 XP_017916635.1 8 - 9 4180 111
XM_018061151.1 XP_017916640.1 9 - 10 4203 111

View Full Product Details