Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102176354
Entrez Gene ID
102176354
Gene Name
haptoglobin
Gene Aliases
HP
Chromosome Location
Chr.18: 40244373 - 40250851 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692202.3 XP_005692259.1 5 - 6 452 106
GenBank mRNA JO420428.2 - 2 - 3 1411 106
JO420429.2 - 2 - 3 1411 106

View Full Product Details