Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GATA1
Entrez Gene ID
102174102
Gene Name
GATA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 21030723 - 21033704 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043633.1 XP_017899122.1 2 - 3 646 58
GenBank mRNA JR125473.1 - - 970 58

View Full Product Details