Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RDH5
Entrez Gene ID
102179453
Gene Name
retinol dehydrogenase 5
Gene Aliases
-
Chromosome Location
Chr.5: 56897314 - 56901840 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680398.3 XP_005680455.1 2 - 3 1084 59
XM_013963965.2 XP_013819419.1 2 - 3 1179 59

View Full Product Details