Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD12
Entrez Gene ID
102182326
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
-
Chromosome Location
Chr.14: 36001583 - 36448448 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689134.3 XP_005689191.1 2 - 3 336 66
XM_018058373.1 XP_017913862.1 2 - 3 336 66
XM_018058374.1 XP_017913863.1 2 - 3 336 66

View Full Product Details