Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C25H7orf61
Entrez Gene ID
102182467
Gene Name
chromosome 25 C7orf61 homolog
Gene Aliases
-
Chromosome Location
Chr.25: 36960303 - 36964400 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040778.1 XP_017896267.1 1 - 2 1154 89

View Full Product Details