Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRAS
Entrez Gene ID
102172519
Gene Name
KRAS proto-oncogene, GTPase
Gene Aliases
-
Chromosome Location
Chr.5: 83368042 - 83412229 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048388.1 XP_017903877.1 2 - 3 306 109
XM_018048389.1 XP_017903878.1 2 - 3 212 109
XM_018048390.1 XP_017903879.1 2 - 3 306 109
XM_018048391.1 XP_017903880.1 2 - 3 306 109

View Full Product Details