Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTN5
Entrez Gene ID
102178563
Gene Name
netrin 5
Gene Aliases
-
Chromosome Location
Chr.18: 56321270 - 56326459 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062731.1 XP_017918220.1 3 - 4 989 105

View Full Product Details