Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SHISA6
Entrez Gene ID
102175241
Gene Name
shisa family member 6
Gene Aliases
-
Chromosome Location
Chr.19: 29775390 - 30036366 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064666.1 XP_017920155.1 1 - 2 676 58
GenBank mRNA JO622743.2 - 1 - 2 798 58

View Full Product Details