Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A4
Entrez Gene ID
102190114
Gene Name
solute carrier family 10 member 4
Gene Aliases
-
Chromosome Location
Chr.6: 67706868 - 67712918 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681596.3 XP_005681653.3 1 - 2 728 57
GenBank mRNA JO616676.2 - 1 - 2 302 57

View Full Product Details