Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL55
Entrez Gene ID
102184707
Gene Name
mitochondrial ribosomal protein L55
Gene Aliases
-
Chromosome Location
Chr.7: 105821720 - 105825579 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682073.3 XP_005682130.1 2 - 3 439 94
XM_018051590.1 XP_017907079.1 2 - 3 436 94
XM_018051591.1 XP_017907080.1 2 - 3 264 94
XM_018051592.1 XP_017907081.1 2 - 3 261 94
GenBank mRNA JO638584.2 - 2 - 3 539 94
KA329441.1 - 2 - 3 262 94

View Full Product Details