Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGT
Entrez Gene ID
102187651
Gene Name
angiotensinogen
Gene Aliases
-
Chromosome Location
Chr.28: 84114 - 95812 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699373.3 XP_005699430.1 4 - 5 1377 70
GenBank mRNA JO421743.2 - 4 - 5 1369 70
JO631757.2 - 1 - 2 678 70

View Full Product Details