The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018043911.1 | XP_017899400.1 | 12 - 13 | 2457 | 111 |
XM_018043912.1 | XP_017899401.1 | 11 - 12 | 2180 | 111 | |
XM_018043913.1 | XP_017899402.1 | 11 - 12 | 2304 | 111 | |
XM_018043915.1 | XP_017899404.1 | 12 - 13 | 2397 | 111 |