Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCLAF3
Entrez Gene ID
102186200
Gene Name
BCLAF1 and THRAP3 family member 3
Gene Aliases
CXHXorf23
Chromosome Location
Chr.X|NW_017189516: 50040370 - 50096850 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043911.1 XP_017899400.1 9 - 10 2089 103
XM_018043912.1 XP_017899401.1 8 - 9 1812 103
XM_018043913.1 XP_017899402.1 8 - 9 1936 103
XM_018043914.1 XP_017899403.1 9 - 10 2089 103
XM_018043915.1 XP_017899404.1 9 - 10 2029 103
XM_018043916.1 XP_017899405.1 9 - 10 2089 103

View Full Product Details