Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
HES4
Entrez Gene ID
106502961
Gene Name
hes family bHLH transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.16: 49971847 - 49975771 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060566.1 XP_017916055.1 2 - 3 1416 108
GenBank mRNA JT709278.1 - 2 - 3 218 108

View Full Product Details