Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- RPL36
- Entrez Gene ID
-
102169094
- Gene Name
- ribosomal protein L36
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 91913894 - 91915408 on Build ARS1
- Gene Symbol
- LONP1
- Entrez Gene ID
-
102182026
- Gene Name
- lon peptidase 1, mitochondrial
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 91915357 - 91933886 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KA350101.1 |
- |
4 - 4 |
364 |
99 |
View Full Product Details