Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUP62
Entrez Gene ID
102187158
Gene Name
nucleoporin 62
Gene Aliases
-
Chromosome Location
Chr.18: 57255749 - 57278155 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062827.1 XP_017918316.1 2 - 2 1907 97
XM_018062828.1 XP_017918317.1 2 - 2 1777 97
XM_018062829.1 XP_017918318.1 2 - 2 1790 97
XM_018062830.1 XP_017918319.1 2 - 2 2415 97

Gene Symbol
ATF5
Entrez Gene ID
102187428
Gene Name
activating transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.18: 57277310 - 57282016 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JT679118.1 - 2 - 2 162 97

View Full Product Details