Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXB6
Entrez Gene ID
102169050
Gene Name
homeobox B6
Gene Aliases
-
Chromosome Location
Chr.19: 37523508 - 37530777 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064934.1 XP_017920423.1 2 - 2 5542 126

View Full Product Details