Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp1b1
Entrez Gene ID
100135483
Gene Name
ATPase Na+/K+ transporting subunit beta 1
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176380: 6166682 - 6187025 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172917.1 NP_001166388.1 2 - 3 484 100
GenBank mRNA AB038514.1 - 2 - 3 484 100

View Full Product Details