Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nqo1
Entrez Gene ID
100135582
Gene Name
NAD(P)H quinone dehydrogenase 1
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176397: 6955543 - 6968929 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172986.1 NP_001166457.1 4 - 5 481 69
XM_005004596.3 XP_005004653.1 4 - 5 506 69
GenBank mRNA AB055403.1 - 4 - 5 481 69

View Full Product Details