Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otos
Entrez Gene ID
100135590
Gene Name
otospiralin
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176406: 23698131 - 23699507 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172993.1 NP_001166464.1 2 - 3 183 75
GenBank mRNA AY062255.1 - 2 - 3 183 75

View Full Product Details