Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mug1
Entrez Gene ID
100216363
Gene Name
murinoglobulin 1
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176391: 1249781 - 1334355 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173061.1 NP_001166532.1 25 - 26 3547 86
GenBank mRNA D84339.1 - 25 - 26 3547 86

View Full Product Details