Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brs3
Entrez Gene ID
100135488
Gene Name
bombesin receptor subtype 3
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176340: 5789417 - 5807686 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172921.1 NP_001166392.1 3 - 4 1114 92
GenBank mRNA X67126.1 - 3 - 4 1114 92

View Full Product Details