Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cryba4
Entrez Gene ID
100306973
Gene Name
crystallin beta A4
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176415: 16274627 - 16280003 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173077.1 NP_001166548.1 3 - 4 188 87
XM_013141518.2 XP_012996972.1 5 - 6 667 87
GenBank mRNA EB371719.1 - 2 - 3 185 87
EF666994.1 - 3 - 4 188 87

View Full Product Details