Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd8b
Entrez Gene ID
100379572
Gene Name
CD8b molecule
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176382: 4120960 - 4136886 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172877.1 NP_001166348.1 3 - 4 551 87
GenBank mRNA AY303774.1 - 3 - 4 551 87
AY303775.1 - 3 - 4 505 87

View Full Product Details