Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpinc1
Entrez Gene ID
100379601
Gene Name
serpin family C member 1
Gene Aliases
ATIII
Chromosome Location
Chr.Un|NT_176407: 9998033 - 10008927 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173401.1 NP_001166872.1 5 - 6 1132 98

View Full Product Details