Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk6
Entrez Gene ID
100379284
Gene Name
potassium two pore domain channel subfamily K member 6
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176367: 11962822 - 11970442 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172763.1 NP_001166234.1 1 - 2 321 81
GenBank mRNA AY075097.1 - 1 - 2 321 81

View Full Product Details