Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DNApol-eta
- Entrez Gene ID
-
40438
- Gene Name
- DNA polymerase eta
- Gene Aliases
-
AAF51794, CG7143, DNA pol eta, DNA pol-eta, DNApoleta, Dmel\CG7143, Dmel_CG7143, Dmpol eta, POLH, dPoleta, drad30A, pol eta, pol h
- Chromosome Location
-
Chr.3L: 21938634 - 21941812 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037141
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141114.3 |
NP_649371.2 |
2 - 3 |
973 |
65 |
View Full Product Details