Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inv
Entrez Gene ID
36239
Gene Name
invected
Gene Aliases
CG17835, Dmel\CG17835, Dmel_CG17835, Engrailed/Invected, INV, IV, Inv, er, in
Chromosome Location
Chr.2R: 11474465 - 11509695 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001269
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078975.4 NP_523699.3 3 - 4 1726 62
NM_165838.3 NP_725056.2 3 - 4 1726 62
NM_165839.3 NP_725057.2 4 - 5 2869 62
GenBank mRNA BT016097.1 - 3 - 4 1617 62

View Full Product Details