Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dp
Entrez Gene ID
36461
Gene Name
DP transcription factor
Gene Aliases
49Fk, CG4654, DDP1, DP, DP1, DmDP, Dmel\CG4654, Dmel_CG4654, Dp1, TFPD2, dDP, dDP1, dDp, dp, l(2)49Fk, l(2)vr10, vr10
Chromosome Location
Chr.2R: 13222433 - 13228202 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011763
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274020.1 NP_001260949.1 2 - 3 231 62
NM_057691.4 NP_477039.1 2 - 3 841 62
NM_165974.3 NP_725267.1 1 - 2 306 62
GenBank mRNA AY069526.1 - 2 - 3 790 62
BT056266.1 - 1 - 2 304 62

View Full Product Details