Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Eip75B
- Entrez Gene ID
-
39999
- Gene Name
- Ecdysone-induced protein 75B
- Gene Aliases
-
57B, 75B, BcDNA:GM02640, CG8127, DmE75, DmE75A, DmE75B, Dmel\CG8127, Dmel_CG8127, E75, E75-C, E75A, E75B, E75C, EP1121b, EiP75, Eip75, Eip75 A, Eip75A, Mgn00274, NR1D3, Rev-Erb, anon-WO0172774.31, anon-WO0172774.32, dE75, eip75B, l(3)07041, l(3)j11A6, l(3)j12E8, l(3)j3A6, l(3)j5E1, l(3)neo25
- Chromosome Location
-
Chr.3L: 17950953 - 18064696 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000568
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168756.2 |
NP_730322.1 |
1 - 2 |
1395 |
118 |
View Full Product Details