Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Papss
Entrez Gene ID
40167
Gene Name
PAPS synthetase
Gene Aliases
CG8363, DmPAPSS, Dmel\CG8363, Dmel_CG8363, PAPS, PAPSS, Paps, l(3)76BDn, paps, papss
Chromosome Location
Chr.3L: 19845293 - 19860925 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020389
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043152.2 NP_001036617.1 4 - 5 2204 67
NM_001275143.1 NP_001262072.1 4 - 5 1907 67
NM_001275144.1 NP_001262073.1 4 - 5 2161 67
NM_079447.4 NP_524171.2 3 - 4 1871 67
NM_168821.3 NP_730457.1 4 - 5 1892 67
NM_168822.3 NP_730458.1 4 - 5 1869 67
NM_168823.3 NP_730459.1 3 - 4 1975 67
NM_168824.3 NP_730460.1 3 - 4 1932 67
GenBank mRNA AY051724.1 - 4 - 5 1856 67
BT100002.1 - 3 - 4 1868 67

View Full Product Details