Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pitslre
Entrez Gene ID
40292
Gene Name
CG4268-PA
Gene Aliases
CG4268, Dmel\CG4268, Dmel_CG4268, Gat, Gta, PITSLRE, PitsIre, cdk11, group 3, pitslre
Chromosome Location
Chr.3L: 20771528 - 20776931 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016696
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259922.2 NP_001246851.1 2 - 3 274 63
NM_001259923.2 NP_001246852.1 1 - 2 333 63
GenBank mRNA BT120362.1 - 2 - 3 236 63

View Full Product Details