Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egr
Entrez Gene ID
36054
Gene Name
eiger
Gene Aliases
BcDNA:RH51659, CG12919, Dmel\CG12919, Dmel_CG12919, Ect1, Egr, Eiger, Regg1, TNF, TNF-alpha, TNFalpha, darth, dt1, regg1
Chromosome Location
Chr.2R: 10078366 - 10084210 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033483
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165735.4 NP_724878.2 4 - 5 1378 89
NM_206069.3 NP_995791.1 4 - 5 1378 89
GenBank mRNA AB073865.1 - 4 - 5 1158 89
AF149799.1 - 4 - 5 1356 89
AF521176.1 - 4 - 5 1372 89
AY115551.1 - 4 - 5 732 89
BT021371.1 - 4 - 5 1356 89
BT072996.1 - 4 - 5 1379 89

View Full Product Details