Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sema2a
Entrez Gene ID
36846
Gene Name
Semaphorin 2a
Gene Aliases
112/40, BcDNA:LD09561, CG4700, CT14922, D-Sema II, D-Sema-II, D-Sema2a, D-SemaII, D-semaII, Dmel\CG4700, Dmel_CG4700, Dsema-II, Sema-2a, SemaII, dSema-II, l(2)03021, sema 2a, sema II, sema-2a, sema-II, sema2a, semaII
Chromosome Location
Chr.2R: 16500178 - 16533459 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011260
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058159.5 NP_477507.1 9 - 10 1490 77
NM_134303.4 NP_599130.1 9 - 10 1391 77
NM_166178.3 NP_725586.1 9 - 10 1427 77
NM_206134.2 NP_995856.2 9 - 10 1350 77
GenBank mRNA AY095026.1 - 9 - 10 1420 77
AY128437.1 - 9 - 10 1335 77

View Full Product Details